missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG2b Heavy Chain Secondary Antibody, DyLight 550, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB7518R
This item is not returnable.
View return policy
Description
IgG2b Heavy Chain Polyclonal antibody specifically detects IgG2b Heavy Chain in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| IgG2b Heavy Chain | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Mouse IgG2b | |
| 1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction