missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat F(ab')2 Anti-mouse IgM-Fluorescein Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems F0118
This item is not returnable.
View return policy
Description
IgM Polyclonal specifically detects IgM in Mouse samples. It is validated for Flow Cytometry.
Specifications
| IgM | |
| Polyclonal | |
| Flow Cytometry | |
| IGHM | |
| Mouse IgM heavy and light chains | |
| 100 Tests | |
| Secondary | |
| Mouse IgM specific IgG was purified by goat IgG affinity chromatography and then adsorbed to eliminate human cross reactivity. The IgG fraction was digested with pepsin to generate F(ab')2 fragments which have a reduced ability to interact with Fc receptors expressed on a variety of cells. | |
| Store the unopened product at 2 - 8degrees C. Do not use past expiration date. Protect from light. |
| Flow Cytometry | |
| Fluorescein | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| 3507 | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction