missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgM (H+L) Secondary Antibody, DyLight 755, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-75203IR
This item is not returnable.
View return policy
Description
IgM (H+L) Polyclonal antibody specifically detects IgM (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion, Immunoassay
Specifications
| IgM (H+L) | |
| Polyclonal | |
| Western Blot : 1:100-1:2000, ELISA : 1:100-1:2000, Immunohistochemistry : 1:10 - 1:500, Immunoprecipitation : 1:10 - 1:500, Immunodiffusion, Immunoassay | |
| Goat | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Purified Mouse IgM, whole molecule | |
| 0.5 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction