missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgM (H+L) Secondary Antibody, Janelia Fluor 549, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-75203JF549
This item is not returnable.
View return policy
Description
IgM (H+L) Polyclonal antibody specifically detects IgM (H+L) in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| IgM (H+L) | |
| Polyclonal | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Goat | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Purified Mouse IgM, whole molecule | |
| 0.5 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction