missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgM (H+L) Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-75203-0.5mg
This item is not returnable.
View return policy
Description
IgM (H+L) Polyclonal specifically detects IgM (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion, Immunoassay.
Specifications
| IgM (H+L) | |
| Polyclonal | |
| Unconjugated | |
| Purified Mouse IgM (H&L) | |
| 0.5 mg | |
| Secondary | |
| Mouse | |
| IgM |
| Immunoassay | |
| 4.5 mg/mL | |
| Goat | |
| Affinity purified | |
| RUO | |
| Based on IEP, this antibody reacts with heavy mu chains on mouse IgM and light chains on all mouse immunoglobulins | |
| Store at 4°C. After dilution, add an equal volume of glycerol to prevent loss of enzymatic activity and store at -20°C. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction