missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgM (Heavy chain) Cross-Adsorbed Secondary Antibody, Biotin, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen M31515
Additional Details : Weight : 0.50000kg
Description
For this goat anti-mouse IgM (Heavy chain) antibody, it is reccommended to use a 1:10,000 working dilution for optimal performance.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Mouse IgM (Heavy chain) Cross-Adsorbed | |
Polyclonal | |
Goat | |
Purified | |
RUO | |
Mouse | |
Liquid |
Immunofluorescence, Western Blot | |
Biotin | |
Mouse Mu immunonglobin | |
1 mL | |
Secondary | |
4° C | |
IgG |