missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgM Heavy Chain Secondary Antibody, DyLight 550, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-75189R
This item is not returnable.
View return policy
Description
IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoassay
Specifications
| IgM Heavy Chain | |
| Polyclonal | |
| Western Blot : 1:100-1:2000, ELISA : 1:100-1:2000, Immunohistochemistry : 1:10 - 1:500, Immunoassay | |
| Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain | |
| Purified Mouse IgM, Mu chain | |
| 0.5 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| 3507 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunoassay | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Goat | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction