missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgM Heavy Chain Secondary Antibody, DyLight 800, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-72884
This item is not returnable.
View return policy
Description
IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Mouse samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, ELISA.
Specifications
| IgM Heavy Chain | |
| Polyclonal | |
| DyLight 800 | |
| Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| No reaction was observed against other mouse heavy or light chain proteins. | |
| Mouse | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:5000, Fluorophore-linked immunosorbent assay 1:20000 | |
| IGHM | |
| Mouse IgM u chain | |
| 0.1 mg | |
| Immunology | |
| 3507 | |
| Reconstitute with 100 ul deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction