Learn More
MTR, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of MTR.
Brand: Abnova PAB6022.100ug
Description
MTR encodes the enzyme 5-methyltetrahydrofolate-homocysteine methyltransferase. This enzyme, also known as cobalamin-dependent methionine synthase, catalyzes the final step in methionine biosynthesis. Mutations in MTR have been identified as the underlying cause of methylcobalamin deficiency complementation group G. [provided by RefSeq
Sequence: C-VEKWLGPILGYDTDSpecifications
| MTR | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)(3-5 ug/mL) ELISA (1:32000) Western blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| MTR | |
| MTR | |
| A synthetic peptide corresponding to human MTR. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of MTR. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| FLJ33168/FLJ43216/FLJ45386/MS | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 4548 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only