Learn More
NEUROD2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of NEUROD2.
Brand: Abnova PAB6948.100ug
Description
This gene encodes a member of the neuroD family of neurogenic basic helix-loop-helix (bHLH) proteins. Expression of this gene can induce transcription from neuron-specific promoters, such as the GAP-43 promoter, which contain a specific DNA sequence known as an E-box. The product of the human gene can induce neurogenic differentiation in non-neuronal cells in Xenopus embryos, and is thought to play a role in the determination and maintenance of neuronal cell fates. [provided by RefSeq
Sequence: C-DMHLHHDRGPMYEESpecifications
| NEUROD2 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) The optimal working dilution should be determined by the end user. | |
| NEUROD2 | |
| NEUROD2 | |
| A synthetic peptide corresponding to human NEUROD2. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of NEUROD2. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| MGC26304/NDRF/bHLHa1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 4761 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only