missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza B, Goat, Biotin, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against yamagata strain of Influenza B.
Brand: Abnova PAB14282.1mL
This item is not returnable.
View return policy
Specifications
| nfluenza B | |
| Polyclonal | |
| Goat polyclonal antibody raised against yamagata strain of Influenza B. | |
| In 10mM PBS, pH 7.2 (0.09% sodium azide) | |
| Influenza B, strain Yamagata. | |
| RUO | |
| Purified virions. Does not cross react with influenza A virus, Parainfluenza virus (types 1-3), Respiratory Syncytial virus or Adenovirus. Negative vs monkey kidney cells by indirect immunofluorescence. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunofluorescence, Western Blot | |
| Biotin | |
| The optimal working dilution should be determined by the end user. | |
| Goat | |
| 1 mL | |
| Primary | |
| Influenza virus | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction