missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKD2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of NKD2.
Brand: Abnova PAB7403.100ug
This item is not returnable.
View return policy
Description
In the mouse, Nkd is a Dishevelled (see DVL1; MIM 601365)-binding protein that functions as a negative regulator of the Wnt (see WNT1; MIM 164820)-beta-catenin (see MIM 116806)-Tcf (see MIM 602272) signaling pathway.[supplied by OMIM
Sequence: C-HKRYRQKGREGHSSpecifications
| NKD2 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) The optimal working dilution should be determined by the end user. | |
| NKD2 | |
| NKD2 | |
| A synthetic peptide corresponding to human NKD2. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of NKD2. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| Naked2 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 85409 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction