Learn More
NONO, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of NONO.
Brand: Abnova PAB7027.100ug
Description
This gene encodes an RNA-binding protein which plays various roles in the nucleus, including transcriptional regulation and RNA splicing. A rearrangement between this gene and the transcription factor E3 gene has been observed in papillary renal cell carcinoma. Alternatively spliced transcript variants have been described. Pseudogenes exist on Chromosomes 2 and 16. [provided by RefSeq
Sequence: C-NRAAPGAEFAPNKSpecifications
| NONO | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) Western Blot (0.1-0.3 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3-6 ug/mL) The optimal working dilution should be determined by the end user. | |
| NONO | |
| NONO | |
| A synthetic peptide corresponding to C-terminus of human NONO. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of NONO. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NMT55/NRB54/P54/P54NRB | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 4841 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only