Learn More
NR1I2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of NR1I2.
Brand: Abnova PAB11612.100ug
Description
This gene product belongs to the nuclear receptor superfamily, members of which are transcription factors characterized by a ligand-binding domain and a DNA-binding domain. The encoded protein is a transcriptional regulator of the cytochrome P450 gene CYP3A4, binding to the response element of the CYP3A4 promoter as a heterodimer with the 9-cis retinoic acid receptor RXR. It is activated by a range of compounds that induce CYP3A4, including dexamethasone and rifampicin. Several alternatively spliced transcripts encoding different isoforms, some of which use non-AUG (CUG) translation initiation codon, have been described for this gene. Additional transcript variants exist, however, they have not been fully characterized. [provided by RefSeq
Sequence: EQFAITLKSYIECNRSpecifications
| NR1I2 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:4000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| NR1I2 | |
| NR1I2 | |
| A synthetic peptide corresponding to human NR1I2. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of NR1I2. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| BXR/ONR1/PAR/PAR1/PAR2/PARq/PRR/PXR/SAR/SXR | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 8856 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only