Learn More
NUMB, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of NUMB.
Brand: Abnova PAB6102.100ug
Description
The protein encoded by this gene plays a role in the determination of cell fates during development. The encoded protein, whose degradation is induced in a proteasome-dependent manner by MDM2, is a membrane-bound protein that has been shown to associate with EPS15, LNX1, and NOTCH1. Four transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: C-PFSSDLQKTFEIELSpecifications
| NUMB | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western Blot (0.03-0.1 ug/mL) The optimal working dilution should be determined by the end user. | |
| NUMB | |
| NUMB | |
| A synthetic peptide corresponding to C-terminus of human NUMB. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize all reported isoforms (NP_001005743.1, 001005744.1, NP_0.03735, NP_001005745.1). It is also likely to cross-react with Numblike (NUMBL, GeneID: 9253) which has a very similar C-Terminus, although this has not yet been tested experimentally. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of NUMB. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| S171 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 8650 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only