Learn More
NUP107, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of NUP107.
Brand: Abnova PAB19031.100ug
Description
This gene encodes a member of the nucleoporin family. The protein is localized to the nuclear rim and is an essential component of the nuclear pore complex (NPC). All molecules entering or leaving the nucleus either diffuse through or are actively transported by the NPC. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized. [provided by RefSeq
Sequence: C-PLPAEDDNAIREHSpecifications
| NUP107 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:8000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| NUP107 | |
| NUP107 | |
| A synthetic peptide corresponding to amino acids at internal region of human NUP107. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of NUP107. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NUP84 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 57122 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only