Learn More
NUP50, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of NUP50.
Brand: Abnova PAB6369.100ug
Description
The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a member of the FG-repeat containing nucleoporins that functions as a soluble cofactor in importin-alpha:beta-mediated nuclear protein import. Pseudogenes of this gene are found on chromosomes 5, 6, and 14. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: C-ELHKILLEKKDASpecifications
| NUP50 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western Blot (0.2-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2.5-3.8 ug/mL) The optimal working dilution should be determined by the end user. | |
| NUP50 | |
| NUP50 | |
| A synthetic peptide corresponding to C-terminus of human NUP50. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_009103.2; NP_705931.1). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of NUP50. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| MGC39961/NPAP60/NPAP60L | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 10762 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only