Learn More
PCGF3, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PCGF3.
Brand: Abnova PAB6332.100ug
Description
The protein encoded by this gene contains a C3HC4 type RING finger, which is a motif known to be involved in protein-protein interactions. The specific function of this protein has not yet been determined. [provided by RefSeq
Sequence: C-PLLLHYRPKMDLLSpecifications
| PCGF3 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:2000) Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| PCGF3 | |
| PCGF3 | |
| A synthetic peptide corresponding to human PCGF3. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize the 247 aa isoform only (NP_006306), not the 132 aa isoform (NP_694982). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of PCGF3. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| DKFZp686D20235/DONG1/FLJ36550/FLJ43813/MGC129615/MGC40413/RNF3/RNF3A | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 10336 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only