missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphotyrosine, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against Phosphotyrosine.
Brand: Abnova PAB13650.250ug
This item is not returnable.
View return policy
Specifications
| Phosphotyrosine | |
| Polyclonal | |
| Goat polyclonal antibody raised against Phosphotyrosine. | |
| In PBS | |
| Phosphotyrosine. | |
| RUO | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Goat | |
| 250 μg | |
| Primary | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction