Learn More
PLIN1 Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PLIN1.
Brand: Abnova PAB7249.100ug
Description
The protein encoded by this gene coats lipid storage droplets in adipocytes, thereby protecting them until they can be broken down by hormone-sensitive lipase. The encoded protein is the major cAMP-dependent protein kinase substrate in adipocytes and, when unphosphorylated, may play a role in the inhibition of lipolysis. Alternatively spliced transcript variants varying in the 5' UTR, but encoding the same protein, have been found for this gene. [provided by RefSeq]
Sequence: C-PREKPKRRVSDSSpecifications
| PLIN1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Western Blot (0.01 ug/mL) The optimal working dilution should be determined by the end user. | |
| PLIN | |
| PLIN | |
| A synthetic peptide corresponding to human PLIN1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of PLIN1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| PERI | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 5346 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only