missing translation for 'onlineSavingsMsg'
Learn More

pnpla2, Goat, Polyclonal Antibody, Abnova™

Product Code. 16070318
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16070318 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16070318 Supplier Abnova Supplier No. PAB7224.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat polyclonal antibody raised against synthetic peptide of pnpla2.

Sequence: C-NIIEVSKEARKR

Specifications

Antigen pnpla2
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of pnpla2.
Dilution ELISA (1:32000) Western blot (0.5-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene Pnpla2
Gene Alias 0610039C21Rik/1110001C14Rik/Atgl/TTS-2.2
Gene Symbols Pnpla2
Host Species Goat
Immunogen A synthetic peptide corresponding to mouse pnpla2.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66853
Test Specificity This antibody was designed using the Mouse sequence NP_080078.1, which differs by 2 amino acids (from the human sequence NP_065109.1.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.