Learn More
PODXL, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PODXL.
Brand: Abnova PAB7292.100ug
Description
This gene encodes a member of the sialomucin protein family. The encoded protein was originally identified as an important component of glomerular podocytes. Podocytes are highly differentiated epithelial cells with interdigitating foot processes covering the outer aspect of the glomerular basement membrane. Other biological activities of the encoded protein include: binding in a membrane protein complex with Na+/H+ exchanger regulatory factor to intracellular cytoskeletal elements, playing a role in hematopoetic cell differentiation, and being expressed in vascular endothelium cells and binding to L-selectin. [provided by RefSeq]
Sequence: C-DNLTKDDLDEEEDTHSpecifications
| PODXL | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:8000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| PODXL | |
| PODXL | |
| A synthetic peptide corresponding to C-terminus of human PODXL. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_001018121.1 and NP_005388.2). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of PODXL. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| Gp200/MGC138240/PC/PCLP | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 5420 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only