missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Porcine IgG Fc Secondary Antibody, DyLight 650, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB729C
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| IgG Fc | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Pig IgG-Fc Fragment | |
| 1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction