missing translation for 'onlineSavingsMsg'
Learn More

PPARGC1A, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of PPARGC1A.

Brand:  Abnova PAB7302.100ug

Product Code. 16050388

  • € 503.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The protein encoded by this gene is a transcriptional coactivator that regulates the genes involved in energy metabolism. This protein interacts with PPARgamma, which permits the interaction of this protein with multiple transcription factors. This protein can interact with, and regulate the activities of, cAMP response element binding protein (CREB) and nuclear respiratory factors (NRFs). It provides a direct link between external physiological stimuli and the regulation of mitochondrial biogenesis, and is a major factor that regulates muscle fiber type determination. This protein may be also involved in controlling blood pressure, regulating cellular cholesterol homoeostasis, and the development of obesity. [provided by RefSeq

Sequence: C-DGLFDDSEDESDK
Specifications

Specifications

PPARGC1A
Polyclonal
Goat polyclonal antibody raised against synthetic peptide of PPARGC1A.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
LEM6/PGC-1(alpha)/PGC-1v/PGC1/PGC1A/PPARGC1
Goat
Antigen affinity purification
RUO
10891
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
Unconjugated
ELISA (1:32000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
PPARGC1A
PPARGC1A
A synthetic peptide corresponding to internal region of human PPARGC1A.
100 μg
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only