Learn More
PPP2R3A Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised aganist synthetic peptide of PPP2R3A.
Brand: Abnova PAB15655.100ug
Description
Protein phosphatase 2 (formerly named type 2A) is one of the four major Ser/Thr phosphatases and is implicated in the negative control of cell growth and division. Protein phosphatase 2 holoenzymes are heterotrimeric proteins composed of a structural subunit A, a catalytic subunit C, and a regulatory subunit B. The regulatory subunit is encoded by a diverse set of genes that have been grouped into the B/PR55, B'/PR61, and B″/PR72 families. These different regulatory subunits confer distinct enzymatic specificities and intracellular localizations to the holozenzyme. The product of this gene belongs to the B″ family. The B″ family has been further divided into subfamilies. The product of this gene belongs to the beta subfamily of regulatory subunit B″. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: TCSNHEQTLSRSpecifications
| PPP2R3A | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:1000) The optimal working dilution should be determined by the end user. | |
| PPP2R3A | |
| PPP2R3A | |
| A synthetic peptide corresponding to amino acids 721-31 of human PPP2R3A. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised aganist synthetic peptide of PPP2R3A. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| PPP2R3/PR130/PR72 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 5523 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only