missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rabbit IgG Fc Secondary Antibody, PE/Cy5.5, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-60661PECY55
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Fluorophore-linked immunosorbent assay
Specifications
| IgG Fc | |
| Polyclonal | |
| PBS | |
| Goat anti-Rabbit IgG Fc fragment Secondary Antibody was produced by repeated immunization with rabbit IgG f(c) fragment in goat | |
| 500 μg | |
| Adaptive Immunity, Immunology | |
| Rabbit | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Fluorophore-Linked Immunosorbent Assay | |
| PE-Cyanine5.5 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction