missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rabbit IgG (H+L) Secondary Antibody, Allophycocyanin/Cy7, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-79817APCCY7
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay | |
| Goat | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Rabbit | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| Purified Rabbit IgG (H&L) | |
| 0.5 mL | |
| Adaptive Immunity, Immunology | |
| Based on Immunoelectrophoresis, this antibody reacts with heavy (gamma) chains on rabbit IgG and light chains on all rabbit immunoglobulins. Cross Adsorbed: This antibody was pre-adsorbed to reduce cross-reactivity with proteins of non-related species. Minimal cross-reactivity with bovine, human, mouse IgG serum proteins. | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction