Learn More
RAD9A, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of RAD9A.
Brand: Abnova PAB7169.100ug
Description
This gene product is highly similar to Schizosaccharomyces pombe rad9, a cell cycle checkpoint protein required for cell cycle arrest and DNA damage repair in response to DNA damage. This protein is found to possess 3' to 5' exonuclease activity, which may contribute to its role in sensing and repairing DNA damage. It forms a checkpoint protein complex with RAD1 and HUS1. This complex is recruited by checkpoint protein RAD17 to the sites of DNA damage, which is thought to be important for triggering the checkpoint-signaling cascade. Use of alternative polyA sites has been noted for this gene. [provided by RefSeq]
Sequence: C-QGPSPVLAEDSEGESpecifications
| RAD9A | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| RAD9A | |
| RAD9A | |
| A synthetic peptide corresponding to human RAD9A. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of RAD9A. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| RAD9 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 5883 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only