Learn More
Goat anti-Rat IgG1 Secondary Antibody, Janelia Fluor 646, Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NBP2-68483JF646
Product Code. 30228649
  • € 508.00 / 0.50mL
Description
IgG1 Polyclonal antibody specifically detects IgG1 in Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG1
Polyclonal
50mM Sodium Borate
Goat
Affinity purified
RUO
Rat
Purified
Western Blot, Flow Cytometry, ELISA
Janelia Fluor 646
IgG1, Igh-4, RGD1359539, VH7183
Rat IgG1
0.5 mL
Secondary
Store at 4°C in the dark.