missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat IgG2a Heavy Chain Secondary Antibody, DyLight 550, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB7122R
This item is not returnable.
View return policy
Description
IgG2a Heavy Chain Polyclonal antibody specifically detects IgG2a Heavy Chain in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| IgG2a Heavy Chain | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rat IgG2a | |
| 1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction