missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat IgM (H+L) Secondary Antibody, DyLight 405, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-68494V
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgM (H+L) Polyclonal antibody specifically detects IgM (H+L) in Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA
Tekniske data
| IgM (H+L) | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rat IgM | |
| 0.5 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, ELISA | |
| DyLight 405 | |
| Goat | |
| Affinity purified | |
| RUO | |
| Rat | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion