missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat IgM Heavy Chain Secondary Antibody, PerCP, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB7100PCP
This item is not returnable.
View return policy
Description
IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Electron Microscopy
Specifications
| IgM Heavy Chain | |
| Polyclonal | |
| PBS | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Secondary | |
| Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Electron Microscopy | |
| PerCP | |
| Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain | |
| Rat IgM | |
| 1 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| 3507 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction