missing translation for 'onlineSavingsMsg'
Learn More

Red Blood Cells, Goat, Polyclonal Antibody, Abnova™

Product Code. 16233550
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16233550 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16233550 Supplier Abnova Supplier No. PAB15383.2mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat polyclonal antibody raised against Red Blood Cells.

Specifications

Antigen Red Blood Cells
Applications Hemolytic Assays
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against Red Blood Cells.
Dilution Immunohistochemistry (1:50-1:200) Western Blot (1:100-1:500) ELISA (1:400-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM sodium phosphate buffer, 140mM NaCl, pH 7.3 (0.05% sodium azide)
Host Species Goat
Immunogen Native purified from rabbit red blood cells.
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This product is suitable for sensitizing cells in hemolytic complement assays, and for lymphocyte screening procedures.
Target Species Rabbit
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 2mL of deionized water store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.