Learn More
RTN4R, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of RTN4R.
Brand: Abnova PAB6793.100ug
Description
This gene encodes the receptor for reticulon 4, oligodendrocyte myelin glycoprotein and myelin-associated glycoprotein. This receptor mediates axonal growth inhibition and may play a role in regulating axonal regeneration and plasticity in the adult central nervous system. [provided by RefSeq
Sequence: C-LQALPDDTFRDLGNSpecifications
| RTN4R | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| RTN4R | |
| RTN4R | |
| A synthetic peptide corresponding to human RTN4R. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of RTN4R. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NGR/NOGOR | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 65078 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only