missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFTPA1/SFTPA2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SFTPA1/SFTPA2.
Brand: Abnova PAB24551.100ug
This item is not returnable.
View return policy
Description
Sequence: C-HLDEELQATLHDFRSpecifications
| SFTPA1/SFTPA2 | |
| Polyclonal | |
| Goat polyclonal antibody raised against synthetic peptide of SFTPA1/SFTPA2. | |
| In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| SFTPA1 | |
| A synthetic peptide corresponding to amino acids 102-115 at internal region of human SFTPA1/SFTPA2. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize all the reported isoforms of A1 (NP_005402.3; NP_001087239.2; NP_001158117.1; NP_001158118.1) and A2 ( NP_001092138.1), Reported variants represent identical protein: NP_001158116.1, NP_005402.3, NP_001158119.1 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:32000) Western Blot (0.01-0.03 ug/mL) The optimal working dilution should be determined by the end user. | |
| COLEC4/FLJ51913/SFTP1/SP-A/SP-A1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 653509/729238 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction