missing translation for 'onlineSavingsMsg'
Learn More
Learn More
six3b, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of six3b.
Brand: Abnova PAB7337.100ug
This item is not returnable.
View return policy
Description
Sequence: C-NRLQHHGLGQSSLRSpecifications
| six3b | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) The optimal working dilution should be determined by the end user. | |
| six3b | |
| six3b | |
| A synthetic peptide corresponding to zebrafish six3b. | |
| 100 μg | |
| Primary | |
| This antibody is not expected to cross-react with Six3a. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of six3b. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| SIX3/cb347/six3.2/six6 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 30636 | |
| Zebrafish | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction