Learn More
SLC7A11, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SLC7A11.
Brand: Abnova PAB7138.100ug
Description
SLC7A11 is a member of a heteromeric Na(+)-independent anionic amino acid transport system highly specific for cystine and glutamate. In this system, designated system Xc(-), the anionic form of cystine is transported in exchange for glutamate.[supplied by OMIM
Sequence: C-KGQTQNFKDAFSGRDSpecifications
| SLC7A11 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Western Blot (0.02-0.06 ug/mL) The optimal working dilution should be determined by the end user. | |
| SLC7A11 | |
| SLC7A11 | |
| A synthetic peptide corresponding to internal region of human SLC7A11. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of SLC7A11. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| CCBR1/xCT | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 23657 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only