Learn More
SPG20, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SPG20.
Brand: Abnova PAB19701.100ug
Description
This gene encodes a protein containing a MIT (Microtubule Interacting and Trafficking molecule) domain, and is implicated in regulating endosomal trafficking and mitochondria function. The protein localizes to mitochondria and partially co-localizes with microtubules. Stimulation with epidermal growth factor (EGF) results in protein translocation to the plasma membrane, and the protein functions in the degradation and intracellular trafficking of EGF receptor. Multiple alternatively spliced variants, encoding the same protein, have been identified. Mutations associated with this gene cause autosomal recessive spastic paraplegia 20 (Troyer syndrome). [provided by RefSeq
Sequence: C-EASGTDVKQLDQGNKSpecifications
| SPG20 | |
| Polyclonal | |
| Goat polyclonal antibody raised against synthetic peptide of SPG20. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| KIAA0610/SPARTIN/TAHCCP1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 23111 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| ELISA (1:8000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| SPG20 | |
| SPG20 | |
| A synthetic peptide corresponding to internal region of human SPG20. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only