missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDGF1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of TDGF1.
Brand: Abnova PAB18688.100ug
This item is not returnable.
View return policy
Description
Sequence: YGRNCEHDVRKENSpecifications
| TDGF1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| TDGF1 | |
| TDGF1 | |
| A synthetic peptide corresponding to amino acids at internal region of human TDGF1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed) | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of TDGF1. | |
| In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| CR/CRGF/CRIPTO/Cripto-1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 6997 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction