missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trf, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against Trf.
Brand: Abnova PAB15384.2mL
This item is not returnable.
View return policy
Specifications
| Trf | |
| Polyclonal | |
| Goat polyclonal antibody raised against Trf. | |
| In 20mM sodium phosphate buffer, 140mM NaCl, pH 7.3 (0.05% sodium azide) | |
| AI266983/Cd176/HP/MGC102653/Tfn/hpx | |
| Goat | |
| 2 mL | |
| Primary | |
| It gives sharp precipitin lines in immunoelectrophoresis and immunodiffusion techniques. It shows reactivity to mouse transferrin; cross-reactivity to other species may exist. Antibody activity to other serum proteins is not present. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunodiffusion, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Trf | |
| Trf | |
| Mouse Trf. | |
| RUO | |
| 22041 | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction