Learn More
TRIP6, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of TRIP6.
Marca: Abnova PAB18589.100ug
Descrizione
This gene is a member of the zyxin family and encodes a protein with three LIM zinc-binding domains. This protein localizes to focal adhesion sites and along actin stress fibers. Recruitment of this protein to the plasma membrane occurs in a lysophosphatidic acid (LPA)-dependent manner and it regulates LPA-induced cell migration. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. [provided by RefSeq
Sequence: C-SHGVLQHTQGLPSpecifica
| TRIP6 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| TRIP6 | |
| TRIP6 | |
| A synthetic peptide corresponding to amino acids at internal region of human TRIP6. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of TRIP6. | |
| In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| MGC10556/MGC10558/MGC29959/MGC3837/MGC4423/OIP1/ZRP-1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 7205 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only