Learn More
UBE2K, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of UBE2K.
Brand: Abnova PAB6301.100ug
Description
The protein encoded by this gene belongs to the ubiquitin-conjugating enzyme family. This protein interacts with RING finger proteins, and it can ubiquitinate huntingtin, the gene product for Huntington's disease. Known functions for this protein include a role in aggregate formation of expanded polyglutamine proteins and the suppression of apoptosis in polyglutamine diseases, a role in the dislocation of newly synthesized MHC class I heavy chains from the endoplasmic reticulum, and involvement in foam cell formation. Multiple transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq]
Sequence: C-SWDVETATELLLSNSpecifications
| UBE2K | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:2000) Western Blot (1-3 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| UBE2K | |
| UBE2K | |
| A synthetic peptide corresponding to C-terminus of human UBE2K. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of UBE2K. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| DKFZp564C1216/DKFZp686J24237/E2-25K/HIP2/HYPG/LIG | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 3093 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only