Learn More
UCHL1 Goat anti-Human, Rat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of UCHL1.
Brand: Abnova PAB19026.100ug
Description
The protein encoded by this gene belongs to the peptidase C12 family. This enzyme is a thiol protease that hydrolyzes a peptide bond at the C-terminal glycine of ubiquitin. This gene is specifically expressed in the neurons and in cells of the diffuse neuroendocrine system. Mutations in this gene may be associated with Parkinson disease
Sequence: EVRFSAVALCKSpecifications
| UCHL1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| UCHL1 | |
| UCHL1 | |
| A synthetic peptide corresponding to amino acids 211-221 at C-terminus of human UCHL1. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of UCHL1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| PARK5/PGP9.5/Uch-L1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 7345 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only