Learn More
UNC13D Goat anti-Human, Mouse, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of UNC13D.
Brand: Abnova PAB7073.100ug
Description
This gene encodes a protein that is a member of the UNC13 family, containing similar domain structure as other family members but lacking an N-terminal phorbol ester-binding C1 domain present in other Munc13 proteins. The protein appears to play a role in vesicle maturation during exocytosis and is involved in regulation of cytolytic granules secretion. Mutations in this gene are associated with familial hemophagocytic lymphohistiocytosis type 3, a genetically heterogeneous, rare autosomal recessive disorder. [provided by RefSeq
Sequence: C-ETQKHKKDLHPLFDSpecifications
| UNC13D | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Western Blot (0.1 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| UNC13D | |
| UNC13D | |
| A synthetic peptide corresponding to human UNC13D. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of UNC13D. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| FHL3/HLH3/HPLH3/Munc13-4 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 201294 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only