missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP4, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of USP4.
Brand: Abnova PAB6404.100ug
This item is not returnable.
View return policy
Description
Sequence: AEGGGCRERPDAESpecifications
| USP4 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:8000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| USP4 | |
| USP4 | |
| A synthetic peptide corresponding to N-terminus of human USP4. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of USP4. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| MGC149848/MGC149849/UNP/Unph | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 7375 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction