missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yc2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of Yc2.
Brand: Abnova PAB19635.100ug
This item is not returnable.
View return policy
Description
Sequence: C-EMDPGIVDNFPLLKSpecifications
| Yc2 | |
| Polyclonal | |
| Goat polyclonal antibody raised against synthetic peptide of Yc2. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| Yc2 | |
| A synthetic peptide corresponding to internal region of rat Yc2. | |
| 100 μg | |
| Primary | |
| Reported variants represent identical protein: NP_001009920.1, NP_001153211.1. Please note that this antibody is expected to cross-react to the homologous Mouse protein Gsta3. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:32000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Yc2 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 494500 | |
| Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction