missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Znrf1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of Znrf1.
Brand: Abnova PAB27672.100ug
This item is not returnable.
View return policy
Description
Sequence: SDSTYAHGNGYQETSpecifications
| Znrf1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western Blot (1-3ug/ml) Immunohistochemistry (10ug/ml) The optimal working dilution should be determined by the end user. | |
| Znrf1 | |
| Znrf1 | |
| A synthetic peptide corresponding to internal region of mouse Znrf1. | |
| 100 μg | |
| Primary | |
| Canine, Chicken, Human, Mouse, Pig, Rat |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of Znrf1. | |
| In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| B830022L21Rik/MGC101991/Rnf42/Zrfp1/nin283 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 170737 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction