Learn More
Goat F(ab')2 anti-Human IgG Fc Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NBP1-75463-0.5mg
 View more versions of this product
Product Code. 18597122
  • € 187.00 / 0.50mg
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay, Chemiluminescence Immunoassay.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG Fc
Polyclonal
Unconjugated
Goat
Affinity purified
RUO
Based on IEP, this antibody reacts with heavy gamma chains on human IgG. This antibody has been pre-adsorbed against bovine, horse, mouse and rabbit serum proteins, and mouse IgG1.
Store at 4°C. After dilution, add an equal volume of glycerol to prevent loss of enzymatic activity and store at -20°C. Avoid repeated freeze/thaw cycles.
Immunoassay
4.5 mg/mL
O75015, O75015, O75015
Purified Human IgG, Fc fragment
0.5 mg
Secondary