missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat F(ab')2 anti-Human IgG (H+L) Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-74960
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion, Immunoassay
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunodiffusion, Immunoassay | |
| Goat | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion | |
| Unconjugated | |
| 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| This Goat F(ab')2 anti-Human IgG (H+L) Secondary Antibody was developed against purified human IgG (H&L). | |
| 1 mg | |
| Adaptive Immunity, Immunology | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction