Learn More
Goat F(ab')2 anti-Human IgG Heavy Chain Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NBP2-68544
Product Code. 18669818
  • € 469.00 / 0.50mg
Description
IgG Heavy Chain Polyclonal antibody specifically detects IgG Heavy Chain in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG Heavy Chain
Polyclonal
Unconjugated
Immunoglobulin G, ImmunoglobulinG
Human IgG
0.5 mg
Immunology
Human
Purified
Western Blot, ELISA, Immunofluorescence
0.5 mg/mL
Borate buffered saline
Goat
Chromatography
RUO
Secondary
Store at 4°C.